  • Search Results
  • The anti-inflammatory and immunosuppressive effects of glucocorticoids, recent developments and mechanistic insights
The anti-inflammatory and immunosuppressive effects of glucocorticoids, recent developments and mechanistic insights
Molecular and Cellular Endocrinology (2011)
Agnes