  • Search Results
  • Labeling and in vivo visualization of transplanted adipose tissue-derived stem cells with safe cadmium-free aqueous ZnS coating of ZnS-AgInS
Labeling and in vivo visualization of transplanted adipose tissue-derived stem cells with safe cadmium-free aqueous ZnS coating of ZnS-AgInS
Scientific reports (2017-01-07)
Yusuke Ogihara, Hiroshi Yukawa, Tatsuya Kameyama, Hiroyasu Nishi, Daisuke Onoshima, Tetsuya Ishikawa, Tsukasa Torimoto, Yoshinobu Baba
ABSTRACT
The facile synthesis of ZnS-AgInS
Sigma-Aldrich
Insulin from bovine pancreas, ≥25 USP units/mg (HPLC), powder