MilliporeSigma
  • A French crop-exposure matrix for use in epidemiological studies on pesticides: PESTIMAT.
A French crop-exposure matrix for use in epidemiological studies on pesticides: PESTIMAT.
Journal of exposure science & environmental epidemiology (2015-12-24)
Isabelle Baldi, Camille Carles, Audrey Blanc-Lapierre, Pascale Fabbro-Peray, Michel Druet-Cabanac, Elisa Boutet-Robinet, Jean-Marc Soulat, Ghislaine Bouvier, Pierre Lebailly
ABSTRACT
Pesticide exposure assessment is a key methodological issue for epidemiological studies. The history of pesticide has proven difficult to obtain from individuals' report because of the wide range of active ingredients (AIs). We developed a crop-exposure matrix, which intends to reconstitute parameters of pesticide exposure in France since 1950. PESTIMAT is composed of tables crossing crops and AIs by year and providing the following metrics: (1) probability (proportion of farmers having used the AIs); (2) frequency (number of treatment days); and (3) intensity (application rate of the AIs in kg/ha). Metrics were obtained by the combination of six sources: (i) registration information from the Agriculture Ministry; (ii) information from agricultural bodies on products marketed; (iii) agricultural recommendations by the Plant Health Protection body; (iv) treatment calendars provided by farmers; (v) data from associations of farmers; and (vi) data from the industry. To date, 529 AIs usable between 1950 and 2010 are included in PESTIMAT: 160 fungicides; 160 herbicides; and 209 insecticides. When combined with duration and determinants of intensity, the metrics in PESTIMAT will make it possible to calculate exposure scores and to search for dose-effect relationships, an important criterion for causality judgment in epidemiology.
Product Description
Supelco
Myclobutanil, PESTANAL®, analytical standard
Supelco
Thiacloprid, PESTANAL®, analytical standard
Supelco
EPTC, PESTANAL®, analytical standard
Supelco
Diflubenzuron, PESTANAL®, analytical standard
Supelco
Procymidone, PESTANAL®, analytical standard
Supelco
Bromoxynil, PESTANAL®, analytical standard
Supelco
Bifenox, PESTANAL®, analytical standard
Supelco
N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine, PESTANAL®, analytical standard