  • Search Results
  • In vitro differentiation of fertile sperm from cryopreserved spermatogonia of the endangered endemic cyprinid honmoroko (Gnathopogon caerulescens).
In vitro differentiation of fertile sperm from cryopreserved spermatogonia of the endangered endemic cyprinid honmoroko (Gnathopogon caerulescens).
Scientific reports (2017-02-18)
Shogo Higaki, Manami Shimada, Kazuaki Kawamoto, Takaaki Todo, Toshihiro Kawasaki, Ikuo Tooyama, Yasuhiro Fujioka, Noriyoshi Sakai, Tatsuyuki Takada
ABSTRACT
Many endemic fish species are threatened with extinction. Conservation strategies and the restoration of endemic fish after extinction must therefore be investigated. Although sperm cryopreservation is indispensable for the conservation of endangered fishes, the limited number of mature fish and limited availability (volume and period) of sperm from small endemic fish hinders the optimization and practical use of this material. In this report, we demonstrate the in vitro differentiation of fertile sperm from cryopreserved spermatogonia of juveniles of the endangered small cyprinid honmoroko (Gnathopogon caerulescens), which is endemic to Lake Biwa in Japan. The entire process of spermatogenesis was recapitulated in vitro using cryopreserved spermatogonia of non-spawning adult and juvenile fish. The differentiation of sperm from spermatogonia was captured as a time-lapse video and confirmed by 5-ethynyl-2'-deoxyuridine (EdU) incorporation into sperm. Fertility was demonstrated by artificial insemination. These results suggest that the combination of cryopreservation of spermatogonia and in vitro sperm differentiation will provide a new and promising strategy for the preservation of paternal genetic materials.
Sigma-Aldrich
Mineral oil, light oil, (neat)
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
L-15 Medium (Leibovitz), Without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Proliferating Cell Nuclear Antigen antibody produced in mouse, clone PC 10, ascites fluid
Sigma-Aldrich
11-Ketotestosterone
Sigma-Aldrich
3,4-Dihydro-2H-pyran, 97%
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Cy3 conjugate, Chemicon®, from goat