  • Search Results
  • Osteopontin has a protective role in prostate tumor development in mice.
Osteopontin has a protective role in prostate tumor development in mice.
European journal of immunology (2016-11-05)
Keiko Danzaki, Masashi Kanayama, Oscar Alcazar, Mari L Shinohara
ABSTRACT
Osteopontin (OPN) is a protein, generally considered to play a pro-tumorigenic role, whereas several reports have demonstrated the anti-tumorigenic function of OPN during tumor development. These opposing anti- and pro-tumorigenic functions are not fully understood. Here, we report that host-derived OPN plays an anti-tumorigenic role in the transgenic adenocarcinoma of the mouse prostate (TRAMP) model and a TRAMP tumor transplant model. Tumor suppression mediated by OPN in Rag2
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Insulin from bovine pancreas, ≥25 USP units/mg (HPLC), powder
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%