  • Search Results
  • Biological and mechanical evaluation of a Bio-Hybrid scaffold for autologous valve tissue engineering.
Biological and mechanical evaluation of a Bio-Hybrid scaffold for autologous valve tissue engineering.
Materials science & engineering. C, Materials for biological applications (2017-02-12)
S Jahnavi, U Saravanan, N Arthi, G S Bhuvaneshwar, T V Kumary, S Rajan, R S Verma
ABSTRACT
Major challenge in heart valve tissue engineering for paediatric patients is the development of an autologous valve with regenerative capacity. Hybrid tissue engineering approach is recently gaining popularity to design scaffolds with desired biological and mechanical properties that can remodel post implantation. In this study, we fabricated aligned nanofibrous Bio-Hybrid scaffold made of decellularized bovine pericardium: polycaprolactone-chitosan with optimized polymer thickness to yield the desired biological and mechanical properties. CD44
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology