  • Search Results
  • The serotonin transporter (SLC6A4) is present in B-cell clones of diverse malignant origin: probing a potential anti-tumor target for psychotropics.
The serotonin transporter (SLC6A4) is present in B-cell clones of diverse malignant origin: probing a potential anti-tumor target for psychotropics.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2005-05-05)
Elizabeth J Meredith, Michelle J Holder, Anita Chamba, Anita Challa, Adrian Drake-Lee, Christopher M Bunce, Mark T Drayson, Geoffrey Pilkington, Randy D Blakely, Martin J S Dyer, Nicholas M Barnes, John Gordon
ABSTRACT
Following our previous description of the serotonin transporter (SERT) acting as a conduit to 5-hydroxytryptamine (5-HT)-mediated apoptosis, specifically in Burkitt's lymphoma, we now detail its expression among a broad spectrum of B cell malignancy, while exploring additional SERT substrates for potential therapeutic activity. SERT was readily detected in derived B cell lines with origins as diverse as B cell precursor acute lymphoblastic leukemia, mantle cell lymphoma, diffuse large B cell lymphoma, and multiple myeloma. Concentration and timecourse kinetics for the antiproliferative and proapoptotic activities of the amphetamine derivatives fenfluramine (an appetite suppressant) and 3,4-methylenedioxymethamphetamine (MDMA; "Ecstasy") revealed them as being similar to the endogenous indoleamine. A tricyclic antidepressant, clomipramine, instead mirrored the behavior of the selective serotonin reuptake inhibitor fluoxetine, both being effective in the low micromolar range. A majority of neoplastic clones were sensitive to one or more of the serotonergic compounds. Dysregulated bcl-2 expression, either by t(14;18)(q32;q21) translocation or its introduction as a constitutively active transgene, provided protection from proapoptotic but not antiproliferative outcomes. These data indicate a potential for SERT as a novel anti-tumor target for amphetamine analogs, while evidence is presented that the seemingly more promising antidepressants are likely impacting malignant B cells independently of the transporter itself.
Sigma-Aldrich
Serotonin hydrochloride, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Fluoxetine hydrochloride, solid
Pricing and availability is not currently available.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.