  • Search Results
  • Response surface methodology applied to the generation of casein hydrolysates with antioxidant and dipeptidyl peptidase IV inhibitory properties.
Response surface methodology applied to the generation of casein hydrolysates with antioxidant and dipeptidyl peptidase IV inhibitory properties.
Journal of the science of food and agriculture (2016-06-09)
Alice B Nongonierma, Solène Le Maux, Claire Esteveny, Richard J FitzGerald
ABSTRACT
Hydrolysis parameters affecting the release of dipeptidyl peptidase IV (DPP-IV) inhibitory and antioxidant peptides from milk proteins have not been extensively studied. Therefore, a multifactorial (i.e. pH, temperature and hydrolysis time) composite design was used to optimise the release of bioactive peptides (BAPs) with DPP-IV inhibitory and antioxidant [oxygen radical absorbance capacity (ORAC)] properties from sodium caseinate. Fifteen sodium caseinate hydrolysates (H1-H15) were generated with Protamex Generation of milk protein hydrolysates through multifactorial design approaches may aid in the optimal enzymatic release of BAPs with serum glucose lowering and antioxidant properties. © 2016 Society of Chemical Industry.
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Ile-Pro-Ile, ≥97% (HPLC)
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines