  • Search Results
  • Photosynthetic Trichomes Contain a Specific Rubisco with a Modified pH-Dependent Activity.
Photosynthetic Trichomes Contain a Specific Rubisco with a Modified pH-Dependent Activity.
Plant physiology (2017-03-03)
Raphaëlle Laterre, Mathieu Pottier, Claire Remacle, Marc Boutry
ABSTRACT
Ribulose-1,5-biphosphate carboxylase/oxygenase (Rubisco) is the most abundant enzyme in plants and is responsible for CO
Sigma-Aldrich
Creatine Phosphokinase from rabbit muscle, Type I, salt-free, lyophilized powder, ≥150 units/mg protein
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
α-Glycerophosphate Dehydrogenase-Triosephosphate Isomerase from rabbit muscle, Type III, ammonium sulfate suspension, GDH 75-200 units/mg protein (biuret), TPI 750-2000 units/mg protein
Sigma-Aldrich
D-Ribulose 1,5-bisphosphate sodium salt hydrate, ≥99.0% (TLC)