  • Search Results
  • Circulating fibronectin contributes to mesangial expansion in a murine model of type 1 diabetes.
Circulating fibronectin contributes to mesangial expansion in a murine model of type 1 diabetes.
Kidney international (2017-02-06)
Verena Klemis, Hiba Ghura, Giuseppina Federico, Carina Würfel, Anke Bentmann, Norbert Gretz, Tatsuhiko Miyazaki, Hermann-Joseph Gröne, Inaam A Nakchbandi
ABSTRACT
Fibronectin is ubiquitously expressed in the extracellular matrix, and its accumulation in the glomerular mesangium in diabetic nephropathy is associated with deterioration of renal function in these patients. However, the exact role of fibronectin in the pathogenesis of diabetic nephropathy remains unknown. To clarify this, we administered fluorescent-labeled plasma fibronectin to wild-type mice and found it to accumulate in the mesangium. Using liver-specific conditional-knockout mice to decrease circulating fibronectin, we reduced circulating fibronectin by more than 90%. In streptozotocin-induced diabetes of these knockout mice, the pronounced fall in circulating fibronectin resulted in a decrease in mesangial expansion by 25% and a decline in albuminuria by 30% compared to diabetic control mice. Indeed, the amount of fibronectin in the kidney was reduced, as was the total amount of collagen. In vitro experiments confirmed that matrix accumulation of fibronectin was enhanced by increasing fibronectin only, glucose only, or the combination of both. Thus, circulating fibronectin contributes to mesangial expansion and exacerbation of albuminuria in a murine model of type 1 diabetes.
Sigma-Aldrich
Anti-Collagen Antibody, Type IV, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Fibronectin Antibody, Chemicon®, from rabbit