  • Search Results
  • A bioprosthetic ovary created using 3D printed microporous scaffolds restores ovarian function in sterilized mice.
A bioprosthetic ovary created using 3D printed microporous scaffolds restores ovarian function in sterilized mice.
Nature communications (2017-05-17)
Monica M Laronda, Alexandra L Rutz, Shuo Xiao, Kelly A Whelan, Francesca E Duncan, Eric W Roth, Teresa K Woodruff, Ramille N Shah
ABSTRACT
Emerging additive manufacturing techniques enable investigation of the effects of pore geometry on cell behavior and function. Here, we 3D print microporous hydrogel scaffolds to test how varying pore geometry, accomplished by manipulating the advancing angle between printed layers, affects the survival of ovarian follicles. 30° and 60° scaffolds provide corners that surround follicles on multiple sides while 90° scaffolds have an open porosity that limits follicle-scaffold interaction. As the amount of scaffold interaction increases, follicle spreading is limited and survival increases. Follicle-seeded scaffolds become highly vascularized and ovarian function is fully restored when implanted in surgically sterilized mice. Moreover, pups are born through natural mating and thrive through maternal lactation. These findings present an in vivo functional ovarian implant designed with 3D printing, and indicate that scaffold pore architecture is a critical variable in additively manufactured scaffold design for functional tissue engineering.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
TWEENĀ® 20, viscous liquid, suitable for cell culture
Sigma-Aldrich
Insulin-transferrin-sodium selenite media supplement, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, vial of ~2,500 IU
Sigma-Aldrich
Tritonā„¢ X-100, for molecular biology
Sigma-Aldrich
trans-Androsterone