MilliporeSigma
Rapid Throughput Analysis of GABA
Molecular pharmacology (2017-04-22)
Atefeh Mousavi Nik, Brandon Pressly, Vikrant Singh, Shane Antrobus, Susan Hulsizer, Michael A Rogawski, Heike Wulff, Isaac N Pessah
ABSTRACT
Fluorometric imaging plate reader membrane potential dye (FMP-Red-Dye) is a proprietary tool for basic discovery and high-throughput drug screening for G-protein-coupled receptors and ion channels. We optimized and validated this potentiometric probe to assay functional modulators of heterologous expressed GABA
Sigma-Aldrich
trans-Androsterone
Supelco
Androsterone, VETRANAL®, analytical standard
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
5α-Pregnan-3α-ol-20-one, solid