  • Search Results
  • Ten-eleven translocation 2 interacts with forkhead box O3 and regulates adult neurogenesis.
Ten-eleven translocation 2 interacts with forkhead box O3 and regulates adult neurogenesis.
Nature communications (2017-07-01)
Xuekun Li, Bing Yao, Li Chen, Yunhee Kang, Yujing Li, Ying Cheng, Liping Li, Li Lin, Zhiqin Wang, Mengli Wang, Feng Pan, Qing Dai, Wei Zhang, Hao Wu, Qiang Shu, Zhaohui Qin, Chuan He, Mingjiang Xu, Peng Jin
ABSTRACT
Emerging evidence suggests that active DNA demethylation machinery plays important epigenetic roles in mammalian adult neurogenesis; however, the precise molecular mechanisms and critical functional players of DNA demethylation in this process remain largely unexplored. Ten-eleven translocation (Tet) proteins convert 5-methylcytosine (5mC) to 5-hydroxymethylcytosine (5hmC) and its downstream derivatives. Here we show that 5hmC is elevated during the differentiation of adult neural stem cells (aNSCs), and Tet2 is primarily responsible for modulating 5hmC dynamics. Depletion of Tet2 leads to increased aNSC proliferation and reduced differentiation in vitro and in vivo. Genome-wide transcriptional analyses reveal important epigenetic roles of Tet2 in maintaining the transcriptome landscape related to neurogenesis. Mechanistically, transcription factor forkhead box O3 (Foxo3a) physically interacts with Tet2 and regulates the expression of genes related to aNSC proliferation. These data together establish an important role for the Tet2-Foxo3a axis in epigenetically regulating critical genes in aNSCs during adult neurogenesis.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Millipore
ANTI-FLAG® M1 Agarose Affinity Gel
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Millipore
EZview Red Anti-c-Myc Affinity Gel
Sigma-Aldrich
Anti-Ki-67 Antibody, Chemicon®, from rabbit