  • Search Results
  • Long-term presence of memory B-cells specific for different vaccine components.
Long-term presence of memory B-cells specific for different vaccine components.
Vaccine (2009-10-06)
A M Buisman, C G H de Rond, K Oztürk, H I Ten Hulscher, R S van Binnendijk
ABSTRACT
Vaccination against infectious diseases ideally should provide lifelong immunity, but in many cases waning of antibody titers has been observed over time. In this study we describe the identification of antigen-specific memory B-cells in peripheral blood of persons born between 1940 and 2004 in The Netherlands. Polyclonal stimulation of either PBMCs or purified B-cells induced proliferation and differentiation of B-cells of the memory phenotype (CD19(+)/CD27(+)) into antibody secreting cells (ASC). Memory B-cells against components of bacterial vaccines (Bordetella pertussis and tetanus) as well as viral vaccines (measles and influenza) were thus identified, even in persons with low serum antibody titers. Enrichment of B-cells increased the sensitivity of memory B-cell detection when compared to PBMCs. Low, but significant correlations between numbers of antigen-specific memory B-cells and the corresponding circulating antibody titers were found for the pertussis proteins and measles virus, but not for tetanus. The identification of the numbers and specificities of peripheral memory B-cells and their relationship with circulating antibodies may be very useful to determine the long-term efficacy of vaccination.
Sigma-Aldrich
Anti-Human IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgG (γ-chain specific) antibody produced in goat, affinity isolated antibody, lyophilized powder
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.