  • Search Results
  • Synbiotic loaded chitosan-Ca-alginate microparticles reduces inflammation in the TNBS model of rat colitis.
Synbiotic loaded chitosan-Ca-alginate microparticles reduces inflammation in the TNBS model of rat colitis.
International journal of pharmaceutics (2017-05-30)
Tanja Petreska Ivanovska, Kristina Mladenovska, Zoran Zhivikj, Maja Jurhar Pavlova, Ivica Gjurovski, Trpe Ristoski, Lidija Petrushevska-Tozi
ABSTRACT
New therapeutic strategies against inflammatory bowel disease (IBD) consider the usage of probiotics, prebiotics and synbiotics as beneficial for the intestinal microbial balance. Limitations of such an approach are addressed into difference in survival, persistence, colonization and variable effects among different probiotic strains, lack in understanding of probiotic mechanisms of action, as well the complex etiology of IBD. The anti-inflammatory activity of Lactobacillus casei 01 (L. casei 01) was assessed in trinitrobenzenesulphonic (TNBS) acid model of rat colitis when the probiotic was used alone and/or in combination with oligofructose-enriched inulin (Synergy 1), and as synbiotic (L. casei 01+Synergy 1) loaded chitosan-Ca-alginate microparticles; all suspended in ayran. The results from the probiotic/synbiotic treatments (8.5-8.9log CFU g
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines