  • Search Results
  • Microsphere cytometry to interrogate microenvironment-dependent cell signaling.
Microsphere cytometry to interrogate microenvironment-dependent cell signaling.
Integrative biology : quantitative biosciences from nano to macro (2017-01-20)
Henriette Christie Ertsås, Garry P Nolan, Mark A LaBarge, James B Lorens
ABSTRACT
Microenvironmental cues comprising surface-mediated and soluble factors control cellular signaling mechanisms underlying normal cellular responses that define homeostatic and diseased cell states. In order to measure cell signaling in single adherent cells, we developed a novel microsphere-based flow cytometry approach. Single normal or neoplastic cells were adhered to uniform microspheres that display mimetic-microenvironments comprising surface combinations of extracellular matrix (ECM) in the presence of soluble agonists/antagonists. Temporal signaling responses were measured with fluorophore-conjugated antibodies that recognize response-dependent epitopes by multiparametric flow cytometry. Using this approach we demonstrate that microenvironment-mimetic combinations of growth factors and extracellular matrix proteins generate distinct cellular signal networks that reveal unique cell signatures in normal and patient biopsy-derived neoplastic cells.
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Protamine sulfate salt from salmon, Grade X, amorphous powder
Sigma-Aldrich
Monoclonal Anti-Collagen, Type I antibody produced in mouse, clone COL-1, ascites fluid
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)