  • Search Results
  • Electrochemical immunosensor based on poly (3,4-ethylenedioxythiophene) modified with gold nanoparticle to detect aflatoxin B
Electrochemical immunosensor based on poly (3,4-ethylenedioxythiophene) modified with gold nanoparticle to detect aflatoxin B
Materials science & engineering. C, Materials for biological applications (2017-05-10)
Arati Sharma, A Kumar, Raju Khan
ABSTRACT
Gold nanoparticles (Au-NPs) functionalized Poly (3,4-ethylenedioxythiophene) (PEDOT) bioelectrodes were fabricated layer by layer deposition on ITO electrode for detection of aflatoxin B
Sigma-Aldrich
3,4-Ethylenedioxythiophene, 97%