US
EN
Applications
Products
Services
Support
Home
Search Results
Comparative analysis of essential oils from eight herbal medicines with pungent flavor and cool nature by GC?MS and chemometric resolution methods
Comparative analysis of essential oils from eight herbal medicines with pungent flavor and cool nature by GC?MS and chemometric resolution methods
Journal of Separation Science (2009)
Zhao C, et al.