  • Search Results
  • A prospective evaluation of a standardized strategy for the use of a polymeric everolimus-eluting bioresorbable scaffold in ST-segment elevation myocardial infarction: Rationale and design of the BVS STEMI STRATEGY-IT study.
A prospective evaluation of a standardized strategy for the use of a polymeric everolimus-eluting bioresorbable scaffold in ST-segment elevation myocardial infarction: Rationale and design of the BVS STEMI STRATEGY-IT study.
Catheterization and cardiovascular interventions : official journal of the Society for Cardiac Angiography & Interventions (2016-10-25)
Alfonso Ielasi, Attilio Varricchio, Gianluca Campo, Massimo Leoncini, Bernardo Cortese, Paolo Vicinelli, Salvatore Brugaletta, Fortunato Scotto di Uccio, Azeem Latib, Maurizio Tespili
ABSTRACT
To assess the feasibility and the clinical results following a prespecified bioresorbable vascular scaffold (Absorb BVS) implantation strategy in ST-elevation myocardial infarction (STEMI) patients. Concerns raised about the BVS safety in STEMI setting because a not negligible thrombosis rate was reported within 30 days and 12 months after implantation. Technical procedural issues related to the structural BVS features were advocated as probable causes for the thrombotic events. This is an investigators-owned and -directed, prospective, nonrandomized, single-arm multicenter registry intended to obtain data from 500 consecutive STEMI patients undergoing primary PCI with BVS (1.1 or GT1) following a prespecified implantation protocol. The study is recorded in ClinicalTrials.gov with the identifier: NCT02601781. The primary endpoint is a device-oriented composite end-point (DOCE) of cardiac death, any myocardial infarction clearly attributable to the intervention culprit vessel and ischemic-driven target lesion revascularization within 30 days after the index procedure. The DOCE will be assessed even at 6-month, 1-, 3-, and 5-year follow-up. This will be the first study investigating the feasibility and the early- and long-term clinical impact of a prespecified BVS implantation protocol in thrombotic lesions causing STEMI. Here, we describe the rationale and the design of the study. © 2016 Wiley Periodicals, Inc.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.