  • Search Results
  • Quercetin and related flavonoids conserve their antioxidant properties despite undergoing chemical or enzymatic oxidation.
Quercetin and related flavonoids conserve their antioxidant properties despite undergoing chemical or enzymatic oxidation.
Food chemistry (2017-05-30)
Elías Atala, Jocelyn Fuentes, María José Wehrhahn, Hernán Speisky
ABSTRACT
Oxidation of a phenolic group in quercetin is assumed to compromise its antioxidant properties. To address this assumption, the ROS-scavenging, Folin-Ciocalteau- and Fe-reducing capacities of quercetin and thirteen structurally related flavonoids were assessed and compared with those of mixtures of metabolites resulting from their chemical and enzymatic oxidation. Regardless of the oxidation mode, the metabolites mixtures largely conserved the antioxidant properties of the parent molecules. For quercetin, 95% of its ROS-scavenging and over 77% of its Folin-Ciocalteau- and Fe-reducing capacities were retained. The susceptibility of flavonoids to oxidative disappearance (monitored by HPLC-DAD) and that of the mixtures to retain their antioxidant capacity was favourably influenced by the presence of a catechol (ring-B) and enol (ring C) function. This is the first study to report that mixtures resulting from the oxidation of quercetin and its analogues largely conserve their antioxidant properties.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Galangin, autophagy inducing flavonoid
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.