  • Search Results
  • The mammalian STE20-like kinase 1 (MST1) is a substrate for the apoptosis inhibiting protein kinase CK2.
The mammalian STE20-like kinase 1 (MST1) is a substrate for the apoptosis inhibiting protein kinase CK2.
Cellular signalling (2017-05-11)
Christina Servas, Sandra Kiehlmeier, Julia Hach, Rebecca Gross, Claudia Götz, Mathias Montenarh
ABSTRACT
Apoptosis and the response to cell stress are evolutionary highly conserved mechanisms. Both processes require strict regulation, which is often performed by protein kinases. The mammalian Sterile 20-like kinase 1 (MST1) is a pro-apoptotic protein kinase, which is activated and cleaved by caspases upon the induction of cell stress. Being a phosphoprotein itself, the activity of MST1 is regulated by phosphorylation. Protein kinase CK2 is an anti-apoptotic protein kinase which seems to be involved in the regulation of many different cellular processes including apoptosis. There is increasing evidence that the cleavage of many substrates by caspases is regulated by phosphorylation in the close vicinity of the caspase cleavage sites. One of these kinases, implicated in the phosphorylation of caspase substrates, is protein kinase CK2. Here, we report that serine 320 of the MST1 protein is a novel phosphorylation site for the anti-apoptotic protein kinase CK2. Although serine 320 is in close vicinity to the caspase 3 cleavage site, caspase 3 cleavage of MST1 is not affected by CK2 phosphorylation. Using biochemical approaches, we were able to show that MST1 co-localizes with the CK2 subunits in the pancreatic β-cell line INS-1 and that full-length MST1 and the activated N-terminal fragment of MST1 both interacted with the CK2 subunits in vitro and in vivo. MST1 is a basophilic kinase whereas CK2 is an acidophilic kinase. Thus, binding of these two kinases in the cytosol and in the nucleus opens the door to the phosphorylation of a variety of new substrates.
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Quinalizarin, ≥95% (HPLC)
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Caspase 3 human, ≥90% (SDS-PAGE), recombinant, expressed in E. coli (C-terminal histidine-tagged), buffered aqueous glycerol solution, ≥1.0 units/mg protein
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat