  • Search Results
  • A high resolution atlas of gene expression in the domestic sheep (Ovis aries).
A high resolution atlas of gene expression in the domestic sheep (Ovis aries).
PLoS genetics (2017-09-16)
Emily L Clark, Stephen J Bush, Mary E B McCulloch, Iseabail L Farquhar, Rachel Young, Lucas Lefevre, Clare Pridans, Hiu G Tsang, Chunlei Wu, Cyrus Afrasiabi, Mick Watson, C Bruce Whitelaw, Tom C Freeman, Kim M Summers, Alan L Archibald, David A Hume
ABSTRACT
Sheep are a key source of meat, milk and fibre for the global livestock sector, and an important biomedical model. Global analysis of gene expression across multiple tissues has aided genome annotation and supported functional annotation of mammalian genes. We present a large-scale RNA-Seq dataset representing all the major organ systems from adult sheep and from several juvenile, neonatal and prenatal developmental time points. The Ovis aries reference genome (Oar v3.1) includes 27,504 genes (20,921 protein coding), of which 25,350 (19,921 protein coding) had detectable expression in at least one tissue in the sheep gene expression atlas dataset. Network-based cluster analysis of this dataset grouped genes according to their expression pattern. The principle of 'guilt by association' was used to infer the function of uncharacterised genes from their co-expression with genes of known function. We describe the overall transcriptional signatures present in the sheep gene expression atlas and assign those signatures, where possible, to specific cell populations or pathways. The findings are related to innate immunity by focusing on clusters with an immune signature, and to the advantages of cross-breeding by examining the patterns of genes exhibiting the greatest expression differences between purebred and crossbred animals. This high-resolution gene expression atlas for sheep is, to our knowledge, the largest transcriptomic dataset from any livestock species to date. It provides a resource to improve the annotation of the current reference genome for sheep, presenting a model transcriptome for ruminants and insight into gene, cell and tissue function at multiple developmental stages.
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides (rough strains) from Salmonella enterica serotype minnesota Re 595 (Re mutant)
Sigma-Aldrich
3-Benzidino-6-(4-chlorophenyl)pyridazine, ≥98% (HPLC), solid
Sigma-Aldrich
Sheep Serum, USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red