  • Search Results
  • MOF as an evolutionarily conserved histone crotonyltransferase and transcriptional activation by histone acetyltransferase-deficient and crotonyltransferase-competent CBP/p300.
MOF as an evolutionarily conserved histone crotonyltransferase and transcriptional activation by histone acetyltransferase-deficient and crotonyltransferase-competent CBP/p300.
Cell discovery (2017-06-06)
Xiaoguang Liu, Wei Wei, Yuting Liu, Xueli Yang, Jian Wu, Yang Zhang, Qiao Zhang, Tieliu Shi, James X Du, Yingming Zhao, Ming Lei, Jin-Qiu Zhou, Jiwen Li, Jiemin Wong
ABSTRACT
Recent studies indicate that histones are subjected to various types of acylation including acetylation, propionylation and crotonylation. CBP and p300 have been shown to catalyze multiple types of acylation but are not conserved in evolution, raising the question as to the existence of other enzymes for histone acylation and the functional relationship between well-characterized acetylation and other types of acylation. In this study, we focus on enzymes catalyzing histone crotonylation and demonstrate that among the known histone acetyltransferases, MOF, in addition to CBP and p300, also possesses histone crotonyltransferase (HCT) activity and this activity is conserved in evolution. We provide evidence that CBP and p300 are the major HCTs in mammalian cells. Furthermore, we have generated novel CBP/p300 mutants with deficient histone acetyltransferase but competent HCT activity. These CBP/p300 mutants can substitute the endogenous CBP/p300 to enhance transcriptional activation in the cell, which correlates with enhanced promoter crotonylation and recruitment of DPF2, a selective reader for crotonylated histones. Taken together, we have identified MOF as an evolutionarily conserved HCT and provide first cellular evidence that CBP/p300 can facilitate transcriptional activation through histone acylation other than acetylation, thus supporting an emerging role for the non-acetylation type of histone acylation in transcription and possibly other chromatin-based processes.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Acetyl coenzyme A lithium salt, ≥93% (HPLC)
Sigma-Aldrich
C646, ≥98% (HPLC)