  • Search Results
  • Endothelial Barrier Protein Expression in Biodegradable Polymer Sirolimus-Eluting Versus Durable Polymer Everolimus-Eluting Metallic Stents.
Endothelial Barrier Protein Expression in Biodegradable Polymer Sirolimus-Eluting Versus Durable Polymer Everolimus-Eluting Metallic Stents.
JACC. Cardiovascular interventions (2017-11-06)
Hiroyoshi Mori, Qi Cheng, Christoph Lutter, Samantha Smith, Liang Guo, Matthew Kutyna, Sho Torii, Emanuel Harari, Eduardo Acampado, Michael Joner, Frank D Kolodgie, Renu Virmani, Aloke V Finn
ABSTRACT
This study sought to investigate endothelial coverage and barrier protein expression following stent implantation. Biodegradable polymer drug-eluting stents (BP-DES) have been purported to have biological advantages in vessel healing versus durable polymer DES (DP-DES), although clinical trial data suggest equipoise. Biodegradable polymer-sirolimus-eluting stents (BP-SES), durable polymer-everolimus-eluting stents (DP-EES), and bare-metal stents (BMS) were compared. In the rabbit model (28, 45, and 120 days), stented arteries underwent light microscopic analysis and immunostaining for the presence of vascular endothelium (VE)-cadherin, an endothelial barrier protein, and were subjected to confocal microscopy and scanning electron microscopy. A cell culture study in stented silicone tubes was performed to assess cell proliferation. Light microscopic assessments were similar between BP-SES and DP-EES. BMS showed nearly complete expression of VE-cadherin at 28 days, whereas both DES showed significantly less with results favoring BP-SES versus DP-EES (39% coverage in BP-SES, 22% in DP-EES, 95% in BMS). Endothelial cell morphologic patterns differed according to stent type with BMS showing a spindle-like shape, DP-EES a cobblestone pattern, and BP-SES a shape in between. VE-cadherin-negative areas showed greater surface monocytes regardless of type of stent. Cell proliferation was suppressed in both DES with numerically less suppression in BP-SES versus DP-EES. This is the first study to examine VE-cadherin expression after DES. All DES demonstrated deficient barrier expression relative to BMS with results favoring BP-SES versus DP-EES. These findings may have important implications for the development of neoatherosclerosis in different stent types.
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture