Profiling the transcriptome with RNA SPOTs.
Nature methods (2017-11-14)
Chee-Huat Linus Eng, Sheel Shah, Julian Thomassie, Long Cai
ABSTRACT
Single-molecule FISH (smFISH) has been the gold standard for quantifying individual transcript abundances. Here, we scale up multiplexed smFISH to the transcriptome level and profile 10,212 different mRNAs from mouse fibroblast and embryonic stem cells. This method, called RNA sequential probing of targets (SPOTs), provides an accurate, flexible, and low-cost alternative to sequencing for profiling transcriptomes.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Tris-EDTA buffer solution, BioUltra, for molecular biology, pH 8.0
Sigma-Aldrich
Triton X-100 solution, BioUltra, for molecular biology, ~10% in H2O
Sigma-Aldrich
Mineral oil, BioReagent, for molecular biology, light oil
Sigma-Aldrich
Lithium chloride, for molecular biology, ≥99%
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., for molecular biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Sigma-Aldrich
Pyranose Oxidase from Coriolus sp., recombinant, expressed in E. coli, ≥2.7 units/mg solid
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., mol wt 6,500-10,000
Sigma-Aldrich
Trimethoxysilane, 95%
Sigma-Aldrich
3-(2-Pyridyldithio)propionic acid N-hydroxysuccinimide ester, ≥95%, powder
Sigma-Aldrich
Cyanoborohydride Coupling Buffer