  • Search Results
  • Biochemical and structural characterization of Marinomonas mediterranead-mannose isomerase Marme_2490 phylogenetically distant from known enzymes.
Biochemical and structural characterization of Marinomonas mediterranead-mannose isomerase Marme_2490 phylogenetically distant from known enzymes.
Biochimie (2017-11-07)
Wataru Saburi, Nongluck Jaito, Koji Kato, Yuka Tanaka, Min Yao, Haruhide Mori
ABSTRACT
d-Mannose isomerase (MI) reversibly isomerizes d-mannose to d-fructose, and is attractive for producing d-mannose from inexpensive d-fructose. It belongs to the N-acylglucosamine 2-epimerase (AGE) superfamily along with AGE, cellobiose 2-epimerase (CE), and aldose-ketose isomerase (AKI). In this study, Marinomonas mediterranea Marme_2490, showing low sequence identity with any known enzymes, was found to isomerize d-mannose as its primary substrate. Marme_2490 also isomerized d-lyxose and 4-OH d-mannose derivatives (d-talose and 4-O-monosaccharyl-d-mannose). Its activity for d-lyxose is known in other d-mannose isomerizing enzymes, such as MI and AKI, but we identified, for the first time, its activity for 4-OH d-mannose derivatives. Marme_2490 did not isomerize d-glucose, as known MIs do not, while AKI isomerizes both d-mannose and d-glucose. Thus, Marme_2490 was concluded to be an MI. The initial and equilibrium reaction products were analyzed by NMR to illuminate mechanistic information regarding the Marme_2490 reaction. The analysis of the initial reaction product revealed that β-d-mannose was formed. In the analysis of the equilibrated reaction products in D
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
D-(+)-Talose, ≥99.0% (HPLC)
Sigma-Aldrich
D-Xylulose, ≥95% (HPLC), syrup
Sigma-Aldrich
D-Altrose, ≥97.0% (HPLC)
Sigma-Aldrich
D-Allose, 98%
Sigma-Aldrich
1,5-Anhydro-D-sorbitol, crystalline
Sigma-Aldrich
6-Deoxy-D-glucose