  • Search Results
  • Lysophosphatidylcholine acyltransferase 4 is involved in chondrogenic differentiation of ATDC5 cells.
Lysophosphatidylcholine acyltransferase 4 is involved in chondrogenic differentiation of ATDC5 cells.
Scientific reports (2017-12-03)
Shirou Tabe, Hisako Hikiji, Wataru Ariyoshi, Tomomi Hashidate-Yoshida, Hideo Shindou, Takao Shimizu, Toshinori Okinaga, Yuji Seta, Kazuhiro Tominaga, Tatsuji Nishihara
ABSTRACT
Glycerophospholipids have important structural and functional roles in cells and are the main components of cellular membranes. Glycerophospholipids are formed via the de novo pathway (Kennedy pathway) and are subsequently matured in the remodeling pathway (Lands' cycle). Lands' cycle consists of two steps: deacylation of phospholipids by phospholipases A
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)