  • Search Results
  • Xanthohumol prevents dextran sulfate sodium-induced colitis via inhibition of IKKβ/NF-κB signaling in mice.
Xanthohumol prevents dextran sulfate sodium-induced colitis via inhibition of IKKβ/NF-κB signaling in mice.
Oncotarget (2018-02-09)
Jae-Min Cho, Sun-Mi Yun, Young-Ho Choi, Jinyuk Heo, Nam-Jung Kim, Seok-Ho Kim, Eun-Hee Kim
ABSTRACT
Xanthohumol (XN), a prenylated chalcone isolated from the hop plant, has been reported to exhibit multiple biological functions including anti-inflammation. However, the pharmacological function of XN on colitis remains unknown. In this study, we investigated the anti-inflammatory effect of synthesized XN and molecular mechanism on dextran sulfate sodium (DSS)-induced experimental colitis. XN attenuated the colitis symptoms along with the prevention of colonic lesions after DSS challenge. XN inhibited the production of pro-inflammatory cytokines, oxidative stress and cyclooxygenase-2 expression in DSS-treated mice. Moreover, XN inhibited the phosphorylation of IκBα, the nuclear translocation of NF-κB subunits and the transcriptional activity of NF-κB