  • Search Results
  • Uhrf1 is indispensable for normal limb growth by regulating chondrocyte differentiation through specific gene expression.
Uhrf1 is indispensable for normal limb growth by regulating chondrocyte differentiation through specific gene expression.
Development (Cambridge, England) (2017-11-29)
Michiko Yamashita, Kazuki Inoue, Noritaka Saeki, Maky Ideta-Otsuka, Yuta Yanagihara, Yuichiro Sawada, Iori Sakakibara, Jiwon Lee, Koichi Ichikawa, Yoshiaki Kamei, Tadahiro Iimura, Katsuhide Igarashi, Yasutsugu Takada, Yuuki Imai
ABSTRACT
Transcriptional regulation can be tightly orchestrated by epigenetic regulators. Among these, ubiquitin-like with PHD and RING finger domains 1 (Uhrf1) is reported to have diverse epigenetic functions, including regulation of DNA methylation. However, the physiological functions of Uhrf1 in skeletal tissues remain unclear. Here, we show that limb mesenchymal cell-specific Uhrf1 conditional knockout mice (
Sigma-Aldrich
Toluidine Blue, for microscopy (Hist., Vit.)
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)