  • Search Results
  • Unusually high levels of bisphenol A (BPA) in thermal paper cash register receipts (CRs): development and application of a robust LC-UV method to quantify BPA in CRs.
Unusually high levels of bisphenol A (BPA) in thermal paper cash register receipts (CRs): development and application of a robust LC-UV method to quantify BPA in CRs.
Toxicology mechanisms and methods (2015-05-30)
Sainath Babu, Sannihith N Uppu, Brittany Martin, Ogad A Agu, Rao M Uppu
ABSTRACT
We have developed a simple, reversed-phase high-performance liquid chromatography (RP-HPLC) method for the determination of bisphenol A (BPA) in thermal paper cash register receipts (CRs). The method is suitable for analysis of other types of bisphenols and it involves an overnight extraction of CRs with acetonitrile (AN) at 50 °C followed by the HPLC analysis on a Supelcosil LC18 column (150 × 4.6 mm, particle size: 5 μ) using 50% AN in water as the mobile phase (5 min, isocratic). The composition of AN in the mobile phase changed to 100% over a 10 min period (linear gradient) and then held at 100% AN for 10 min (isocratic). The flow rate was set at 1 mL/min (injection volume: 20 μL) and the eluent was monitored at 234 nm. The authentic BPA eluted with a retention time of 5.9 min and gave a linear detector response in the concentration range of 0.23-50 mg/L. BPA in the CR extracts also eluted with the same retention and had identical absorbance properties as the standard. When CR extracts were co-injected with authentic BPA, they were resolved as a single peak. Further, GC/MS/EI analysis of authentic BPA and the HPLC-purified CR extracts have identical ion chromatograms and fragmentation of the molecular ion (m/z = 228). We have analyzed 170 CRs collected from 62 different vendors including supermarkets, fast food restaurants, gas stations and banking outlets. Almost all cash receipts (n = 168) showed the presence of BPA in the concentration range of 0.45-4.26% (M ± SD, 1.54 ± 0.73%).
Product Description
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-DABS HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm