  • Search Results
  • Acrylamide-functionalized graphene micro-solid-phase extraction coupled to high-performance liquid chromatography for the online analysis of trace monoamine acidic metabolites in biological samples.
Acrylamide-functionalized graphene micro-solid-phase extraction coupled to high-performance liquid chromatography for the online analysis of trace monoamine acidic metabolites in biological samples.
Journal of separation science (2015-02-07)
Xiaoting Yang, Yufei Hu, Gongke Li, Zhuomin Zhang
ABSTRACT
Monoamine acidic metabolites in biological samples are essential biomarkers for the diagnosis of neurological disorders. In this work, acrylamide-functionalized graphene adsorbent was successfully synthesized by a chemical functionalization method and was packed in a homemade polyether ether ketone micro column as a micro-solid-phase extraction unit. This micro-solid-phase extraction unit was directly coupled to high-performance liquid chromatography to form an online system for the separation and analysis of three monoamine acidic metabolites including homovanillic acid, 5-hydroxyindole-3-acetic acid, and 3,4-dihydroxyphenylacetic acid in human urine and plasma. The online system showed high stability, permeability, and adsorption capacity toward target metabolites. The saturated extraction amount of this online system was 213.1, 107.0, and 153.4 ng for homovanillic acid, 5-hydroxyindole-3-acetic acid, and 3,4-dihydroxyphenylacetic acid, respectively. Excellent detection limits were achieved in the range of 0.08-0.25 μg/L with good linearity and reproducibility. It was interesting that three targets in urine and plasma could be actually quantified to be 0.94-3.93 μg/L in plasma and 7.15-19.38 μg/L in urine. Good recoveries were achieved as 84.8-101.4% for urine and 77.8-95.1% for plasma with the intra- and interday relative standard deviations less than 9.3 and 10.3%, respectively. This method shows great potential for online analysis of trace monoamine acidic metabolites in biological samples.
Product Description
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm (PDMS), needle size 24 ga, for use with autosampler
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm (PA), for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm (PDMS), needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm (PDMS), for use with autosampler, needle size 23 ga
Supelco
SPME LC Tips, PDMS/DVB, pkg of 96 ea
Supelco
SPME-OC Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 75 μm (Coating thickness includes 65 μm coating + 10 μm OC (overcoating)), needle size 23 ga, pkg of 3 ea, fiber L 1 cm
Supelco
IonSense® C18 SPE-it Tips, pkg of 96 ea
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm (PDMS), needle size 23 ga, for use with autosampler
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm (PEG), metal alloy fiber, for use with manual holder
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm (PEG), for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (PDMS/DVB), needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME StableFlex fiber assortment kit, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga