• Home
  • Search Results
  • Translocation-positive low-grade fibromyxoid sarcoma: clinicopathologic and molecular analysis of a series expanding the morphologic spectrum and suggesting potential relationship to sclerosing epithelioid fibrosarcoma: a study from the French Sarcoma Group.
Translocation-positive low-grade fibromyxoid sarcoma: clinicopathologic and molecular analysis of a series expanding the morphologic spectrum and suggesting potential relationship to sclerosing epithelioid fibrosarcoma: a study from the French Sarcoma Group.
The American journal of surgical pathology (2007-08-28)
Louis Guillou, Jean Benhattar, Carole Gengler, Gabrielle Gallagher, Dominique Ranchère-Vince, Françoise Collin, Philippe Terrier, Marie-José Terrier-Lacombe, Agnès Leroux, Bernard Marquès, Nicolas de Saint Aubain Somerhausen, Frédérique Keslair, Florence Pedeutour, Jean-Michel Coindre
ABSTRACT
Low-grade fibromyxoid sarcomas (LGFMS) bear either the t(7,16) (q32-34;p11) or t(11,16) (p11;p11) translocations, resulting in FUS-CREB3L2 or FUS-CREB3L1 fusions, respectively. Heretofore, fusion transcripts were mainly detected in frozen tissues, using reverse transcription-polymerase chain reaction. In this study, we aimed to develop a reliable method to detect these in paraffin-embedded tissues, and to examine the clinicopathologic characteristics of a series of translocation-positive LGFMS. Sixty-three neoplasms with typical morphologic features of LGFMS and 66 non-LGFMS tumors selected for their resemblance to LGFMS (LGFMS-like tumors) were examined. RNA of sufficient quality could be extracted from 111/129 (86%) cases (59 LGFMS, 52 non-LGFMS). Of all, 48/59 (sensitivity, 81%) LGFMS contained detectable transcripts (45 FUS-CREB3L2, 3 FUS-CREB3L1). Most relevant clinicopathologic features of fusion-positive LGFMS included predominance in lower extremities (22/48; thigh: 13/48), deep situation (46/48), and occasional presence of unusual histologic features, for example, hypercellular areas (16/48), foci of epithelioid cells (13/48), and giant rosettes (6/48). Most tumors expressed EMA (41/45), at least focally, CD99 (38/41) and bcl-2 (36/41) while being essentially negative for CD34 (2/45), mdm2 (1/41), smooth muscle actin (1/45), S100 protein (0/46), desmin (0/44), h-caldesmon (0/42), keratins (0/44), and CD117 (0/40). Eleven presumed LGFMS were fusion negative. Of all, 7/52 non-LGMFS neoplasms contained FUS-CREB3L2 transcripts, of which 4 had been diagnosed as sclerosing epithelioid fibrosarcoma. In conclusion, FUS-CREB3L1/L2 fusion transcripts can be detected in paraffin-embedded LGFMS in a sensitive manner, using reverse transcription-polymerase chain reaction. Most fusion-positive LGFMS are EMA-positive and CD34/S100/smooth muscle actin negative. The presence of epithelioid cells and fusion transcripts in both LGFMS and a subset of sclerosing epithelioid fibrosarcoma suggest that these neoplasms might be related.