• Home
  • Search Results
  • Correlation of kidney function, volume and imaging findings, and PKHD1 mutations in 73 patients with autosomal recessive polycystic kidney disease.
Correlation of kidney function, volume and imaging findings, and PKHD1 mutations in 73 patients with autosomal recessive polycystic kidney disease.
Clinical journal of the American Society of Nephrology : CJASN (2010-04-24)
Meral Gunay-Aygun, Esperanza Font-Montgomery, Linda Lukose, Maya Tuchman, Jennifer Graf, Joy C Bryant, Robert Kleta, Angelica Garcia, Hailey Edwards, Katie Piwnica-Worms, David Adams, Isa Bernardini, Roxanne E Fischer, Donna Krasnewich, Neal Oden, Alex Ling, Zenaide Quezado, Colleen Zak, Kailash T Daryanani, Baris Turkbey, Peter Choyke, Lisa M Guay-Woodford, William A Gahl
ABSTRACT
Renal function and imaging findings have not been comprehensively and prospectively characterized in a broad age range of patients with molecularly confirmed autosomal recessive polycystic kidney disease (ARPKD). Ninety potential ARPKD patients were examined at the National Institutes of Health Clinical Center. Seventy-three fulfilled clinical diagnostic criteria, had at least one PKHD1 mutation, and were prospectively evaluated using magnetic resonance imaging (MRI), high-resolution ultrasonography (HR-USG), and measures of glomerular and tubular function. Among 31 perinatally symptomatic patients, 25% required renal replacement therapy by age 11 years; among 42 patients who became symptomatic beyond 1 month (nonperinatal), 25% required kidney transplantation by age 32 years. Creatinine clearance (CrCl) for nonperinatal patients (103 +/- 54 ml/min/1.73 m(2)) was greater than for perinatal patients (62 +/- 33) (P = 0.002). Corticomedullary involvement on HR-USG was associated with a significantly worse mean CrCl (61 +/- 32) in comparison with medullary involvement only (131 +/- 46) (P < 0.0001). Among children with enlarged kidneys, volume correlated inversely with function, although with wide variability. Severity of PKHD1 mutations did not determine kidney size or function. In 35% of patients with medullary-only abnormalities, standard ultrasound was normal and the pathology was detectable with HR-USG. In ARPKD, perinatal presentation and corticomedullary involvement are associated with faster progression of kidney disease. Mild ARPKD is best detected by HR-USG. Considerable variability occurs that is not explained by the type of PKHD1 mutation.