Minor histocompatibility antigens.
Current opinion in immunology (1997-11-22)
E Simpson, D Roopenian
ABSTRACT
The existence of transplantation antigens, in addition to those encoded by genes in the MHC, has been known for over half a century. The molecular identification of these additional minor histocompatibility (H) antigens lagged behind that of their MHC counterparts, largely because minor H antigens are recognised by T cells and not by antibodies. In the past year, however, new minor H antigens have been identified at both the genetic and protein level and include Uty, a second novel gene encoding a male-specific epitope in mice, a novel autosomal gene encoding each of the H-13 alleles of mice, and a second male-specific epitope encoded by the SMCY gene.