MilliporeSigma
ASTM D6526: GC Analysis of Impurities in Toluene on SLB®-IL100, 30 m Column
ASTM D6526: GC Analysis of Impurities in Toluene on SLB®-IL100, 30 m Column suitable for GC
Materials
related product
Product No.
Description
Pricing
SLB®-IL100 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SLB-IL100, 30 m x 0.25 mm I.D., 0.20 μm (28884-U)
oven
70 °C
inj. temp.
200 °C
detector
FID, 200 °C
carrier gas
helium, 28 cm/sec
injection
1 μL 40:1 split
liner
4 mm I.D., split type, cup design
sample
toluene, with each aromatic impurity spiked at 0.01% by weight
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany