MilliporeSigma
ASTM D6526: GC Analysis of Impurities in Toluene on SLB®-IL100, 60 m Column
ASTM D6526: GC Analysis of Impurities in Toluene on SLB®-IL100, 60 m Column suitable for GC
Materials
related product
Product No.
Description
Pricing
SLB®-IL100 Capillary GC Column
L × I.D. 60 m × 0.32 mm, df 0.26 μm
CONDITIONS
column
SLB-IL100, 60 m x 0.32 mm I.D., 0.20 μm (28888-U)
oven
70 °C
inj. temp.
150 °C
detector
230 °C
carrier gas
helium, 25 cm/sec
injection
1 μL 40:1 split
sample
toluene spiked with 100 ppm (v/v) of benzene, ethylbenzene, p-xylene, cumene, o-xylene, and dioxane
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany