GC Analysis of Aromatics and Alcohols on SLB®-IL111i, Isothermal Analysis
Materials
related product
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL111i, 60 m x 0.25 mm I.D., 0.20 μm (29884-U)
oven
50 °C (20 min)
inj. temp.
250 °C
detector
FID, 200 °C
carrier gas
helium, 21 cm/sec
injection
1 μL, 100:1 split
liner
4 mm I.D., split type, cup design (2879925-U)
sample
8 analytes at various concentrations in isooctane
Description
Analysis Note
The inert SLB IL111i ionic liquic column produces great peak shapes for alcohols via an isothermal analysis, providing resolution in a mix of aromatics. This is impressive for such a polar column.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany