MilliporeSigma
GC Analysis of Aromatics and Alcohols on SLB®-IL111i, Temperature Programmed Analysis
GC Analysis of Aromatics and Alcohols on SLB®-IL111i, Temperature Programmed Analysis suitable for GC
Materials
related product
Product No.
Description
Pricing
SLB®-IL111i Capillary GC Column
L × I.D. 60 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SLB-IL111i, 60 m x 0.25 mm I.D., 0.20 μm (29884-U)
oven
50 °C (6 min), 15 °C/min to 265 °C (10 min)
inj. temp.
250 °C
detector
FID, 250 °
carrier gas
helium, 21 cm/sec
injection
1 μL, 100:1 split
liner
4 mm I.D., split type, cup design (2879925-U)
sample
8 analytes at various concentrations in isooctane
Description
Analysis Note
The inert SLB IL111i ionic liquic column produces great peak shapes for alcohols via a temperature programmed analysis, providing resolution in a mix of aromatics. This is impressive for such a polar column.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany