MilliporeSigma
GC Analysis of Ionic Liquid Test Mix on SLB®-IL100 (30 m x 0.25 mm I.D., 0.20 μm)
GC Analysis of Ionic Liquid Test Mix on SLB®-IL100 (30 m x 0.25 mm I.D., 0.20 μm) suitable for GC
Materials
related product
Product No.
Description
Pricing
SLB®-IL100 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SLB-IL100, 30 m x 0.25 mm I.D., 0.20 μm (28884-U)
oven
110 °C
inj. temp.
250 °C
detector
FID, 250 °C
detector
helium, 26 cm/sec
injection
1.0 μL 100:1 split
sample
Ionic Liquid Test Mix #1, each analyte at various concentrations in isooctane (N09000)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany