MilliporeSigma
GC Analysis of Ionic Liquid Test Mix on SLB®-IL59 (15 m x 0.10 mm I.D., 0.08 μm)
GC Analysis of Ionic Liquid Test Mix on SLB®-IL59 (15 m x 0.10 mm I.D., 0.08 μm) suitable for GC
Materials
related product
Product No.
Description
Pricing
SLB®-IL59 Capillary GC Column
L × I.D. 15 m × 0.10 mm, df 0.08 μm
CONDITIONS
column
SLB-IL59, 15 m x 0.10 mm I.D., 0.08 μm (28880-U)
oven
90 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
hydrogen, 50 cm/sec
injection
0.5 μL 300:1 split
sample
Ionic Liquid Test Mix #1, each analyte at various concentrations in isooctane (N09000)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany