MilliporeSigma
GC Analysis of Ionic Liquid Test Mix on SLB®-IL76 (15 m x 0.10 mm I.D., 0.08 μm)
GC Analysis of Ionic Liquid Test Mix on SLB®-IL76 (15 m x 0.10 mm I.D., 0.08 μm) suitable for GC
CONDITIONS
column
SLB-IL76, 15 m x 0.10 mm I.D., 0.08 μm (28909-U)
oven
90 °C
inj. temp.
250 °C
detector
FID, 250 °C
injection
0.5 μL 300:1 split
sample
Ionic Liquid Test Mix #1 (N09000), each analyte at various concentrations in isooctane
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany