MilliporeSigma
GC Analysis of Peppermint Essential Oil on SUPELCOWAX 10
GC Analysis of Peppermint Essential Oil on SUPELCOWAX™ 10 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOWAX 10 Capillary GC Column
L × I.D. 60 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SUPELCOWAX 10, 60 m × 0.25 mm I.D., 0.25 μm (24081)
oven
75 °C (4 min.), 4 °C/min. to 200 °C (5 min.)
detector
FID
carrier gas
helium, 25 cm/sec @ 155 °C
injection
0.2 μL, 100:1 split
Description
Legal Information
SUPELCOWAX is a trademark of Sigma-Aldrich Co. LLC