MilliporeSigma
GC Analysis of Phthalate Esters on Equity-5
GC Analysis of Phthalate Esters on Equity™-5 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Equity-5 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
Equity-5, 30 m × 0.25 mm I.D., 0.25 μm (28089-U)
oven
40 °C (1 min), 10 °C/min to 325 °C
inj. temp.
250 °C
detector
FID
carrier gas
helium, 1.3 mL/min constant flow
injection
1 μL, splitless (0.75 min)
liner
4 mm I.D., single taper
sample
50 ng on-column of a custom phthalate ester mix
Description
Legal Information
Equity is a trademark of Sigma-Aldrich Co. LLC