GC Analysis of Semivolatiles in Water on a 5% Phenyl Column after SPME using 7 μm PDMS
Materials
related product
SPME Fiber Assembly
7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hubCONDITIONS
sample/matrix
water spiked with PAHs and phthalates at 50 ppb
SPME fiber
polydimethylsiloxane, 7 μm (57302)
extraction
direct immersion in water, 15 min (rapid stirring)
column
5% Phenyl Polysiloxane, 30 m × 0.25 mm I.D., 0.25 μm (Supelco equivalent, SLB-5ms, 28471-U)
oven
60 °C (3 min), 10 °C/min to 320 °C
inj. temp.
280 °C
detector
MS, scan range m/z = 45-465 at 0.6 sec/scan
carrier gas
helium, 40 cm/sec at 60 °C