MilliporeSigma
GC Analysis of Xylene Isomers on SLB®-IL60
GC Analysis of Xylene Isomers on SLB®-IL60 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-IL60 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SLB-IL60, 30 m x 0.25 mm I.D., 0.20 μm (29505-U)
oven
40 °C (4 min), 8 °C/min to 200 °C (5 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
30 cm/sec constant pressure
injection
1 μL 100:1 split
liner
4 mm I.D., split/splitless type, wool packed single taper FocusLiner design
sample
12 analytes, each at 0.2% (v/v) in pentane
Description
Analysis Note
The xylene isomers are precursors to many chemicals:
  • o-xylene is a precursor for phthalic anhydride
  • m-xylene is a precursor for isophthalic acid
  • p-xylene is a precursor for terephthalic acid and dimethyl terephthalate
The cresol (methylphenol) isomers are also precursors to many chemicals. This chromatogram of a mix of aromatic and methylphenol compounds was generated using an SLB-IL60 ionic liquid column. Its interaction mechanisms allow the separation of all three xylene isomers, and all three cresol isomers.
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany