HPLC Analysis of Antihistamines on Discovery® C8
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C8, 15 cm × 4.6 mm I.D., 5 μm particles (59353-U)
mobile phase
[A] acetonitrile; [B] 25 mM potassium phosphate, pH 2.3; (8:92, A:B)
flow rate
2 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
2 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany