HPLC Analysis of Cardiac Drugs on Ascentis® C8
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis C8, 15 cm x 2.1 mm I.D., 3 μm particles (581402-U)
mobile phase
0.1% ammonium acetate:acetonitrile with 0.1% ammonium acetate, (47:53)
flow rate
0.2 mL/min
column temp.
30 °C
detector
UV, 220 nm
injection
2 μL
sample
as indicated in mobile phase
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany