MilliporeSigma
HPLC Analysis of Chlorhexidine and Robenidine on Ascentis® Phenyl
HPLC Analysis of Chlorhexidine and Robenidine on Ascentis® Phenyl application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Phenyl HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis Phenyl 15 cm x 4.6 mm I.D., 5 μm particles (581616-U)
mobile phase
[A] water; [B] acetonitrile; [C] 500 mM ammonium formate, pH 3.0 with formic acid
gradient
10% C constant; 65% A, 25% B to 0% A, 90% B in 7 min; held for 2 min; to 65% A, 25% B in 0.5 min
flow rate
1.0 mL/min
column temp.
35 °C
detector
UV, 273 nm
injection
10 μL
sample
100 ug/mL each in 70:30, 25 mM ammonium formate pH 3.0:acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany