MilliporeSigma
HPLC Analysis of Nucleosides on TSKgel® Boronate-5PW, Isocratic Mode
HPLC Analysis of Nucleosides on TSKgel® Boronate-5PW, Isocratic Mode application for HPLC
Materials
analytical column
Product No.
Description
Pricing
TSKgel® Boronate-5PW HPLC Column
phase m-aminophenyl boronate, L × I.D. 7.5 cm × 7.5 mm, 10 μm particle size
guard column
Product No.
Description
Pricing
TSKgel® Boronate-5PW HPLC Guardgel Kit
phase m-aminophenyl boronate, L × I.D. 2.5 cm × 6 mm, 20 μm particle size
related product
Product No.
Description
Pricing
TSKgel® Boronate-5PW HPLC Column
phase m-aminophenyl boronate, L × I.D. 5 cm × 5 mm, 10 μm particle size, glass
TSKgel® Boronate-5PW HPLC Guardgel Kit
phase m-aminophenyl boronate, L × I.D. 1 cm × 8 mm, 20 μm particle size, glass
standard
Product No.
Description
Pricing
Guanosine
≥98%
Adenosine
United States Pharmacopeia (USP) Reference Standard
CONDITIONS
column
TSKgel Boronate-5PW, 7.5 cm x 7.5 mm I.D., 10 μm particles (813066)
mobile phase
0.1 M phosphate buffer, pH 8.0
flow rate
1 mL/min
detector
UV, 280 nm
Description
Other Notes
Data provided by Tosoh Bioscience LLC.
Legal Information
TSKgel is a registered trademark of Tosoh Corporation