MilliporeSigma
HPLC Analysis of Nucleotides on SUPELCOSIL LC-18-T
HPLC Analysis of Nucleotides on SUPELCOSIL™ LC-18-T application for HPLC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOSIL LC-18-T HPLC Column
3 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
SUPELCOSIL LC-18-T, 15 cm × 4.6 mm I.D., 3 μm particles (58970-U)
mobile phase
[A] 0.1M potassium phosphate buffer/4mM tertrabutylammonium hydrogen sulfate, pH6.0; [B] A:methanol (70:30), pH 7.2
flow rate
1.5 mL/min
detector
UV, 254 nm
injection
20 μL deionized water (25-530 μM each analyte)
Description
Legal Information
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC